Product Details
- SNP ID
-
rs201313732
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:169400339 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGAACAAATTTCAATATTTTCAT[C/T]TTCATCTTCATGAATAGCCTATACC
- Phenotype
-
MIM: 604588
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NEK1
PubMed Links
Gene Details
- Gene
- NEK1
- Gene Name
- NIMA related kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199397.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1245D |
NP_001186326.1 |
NM_001199398.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1201D |
NP_001186327.1 |
NM_001199399.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1148D |
NP_001186328.1 |
NM_001199400.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1173D |
NP_001186329.1 |
NM_012224.2 |
4229 |
Missense Mutation |
AAT,GAT |
N1217D |
NP_036356.1 |
XM_006714228.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1186D |
XP_006714291.1 |
XM_011532003.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1217D |
XP_011530305.1 |
XM_011532004.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1173D |
XP_011530306.1 |
XM_011532005.2 |
4229 |
Intron |
|
|
XP_011530307.1 |
XM_017008249.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1038D |
XP_016863738.1 |
XM_017008250.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1038D |
XP_016863739.1 |
XM_017008251.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1010D |
XP_016863740.1 |
XM_017008252.1 |
4229 |
Missense Mutation |
AAT,GAT |
N1010D |
XP_016863741.1 |
XM_017008253.1 |
4229 |
Missense Mutation |
AAT,GAT |
N861D |
XP_016863742.1 |
XM_017008254.1 |
4229 |
Missense Mutation |
AAT,GAT |
N793D |
XP_016863743.1 |
View Full Product Details