Product Details

SNP ID
rs201281099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139454417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGGTGGATTTCCGAGAACGAGC[A/G]GTGGAGATTGATGGGGAGCGCATCA
Phenotype
MIM: 605950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB33B PubMed Links

Gene Details

Gene
RAB33B
Gene Name
RAB33B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031296.2 707 Silent Mutation GCA,GCG A74A NP_112586.1
XM_011532299.1 707 Silent Mutation GCA,GCG A122A XP_011530601.1

View Full Product Details