Product Details

SNP ID
rs201401118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48499492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGTTGGTATGATTGCACCATG[A/C]GTAGAGACTCTCTTATTTCCAGATT
Phenotype
MIM: 607520
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FRYL PubMed Links

Gene Details

Gene
FRYL
Gene Name
FRY like transcription coactivator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015030.1 6268 Missense Mutation CGC,CTC R2991L NP_055845.1
XM_005248090.4 6268 Missense Mutation CGC,CTC R3006L XP_005248147.1
XM_005248093.4 6268 Missense Mutation CGC,CTC R2022L XP_005248150.1
XM_011513680.2 6268 Missense Mutation CGC,CTC R3005L XP_011511982.1
XM_011513681.2 6268 Missense Mutation CGC,CTC R3000L XP_011511983.1
XM_011513682.2 6268 Missense Mutation CGC,CTC R2985L XP_011511984.1
XM_011513683.2 6268 Missense Mutation CGC,CTC R3006L XP_011511985.1
XM_011513684.2 6268 Missense Mutation CGC,CTC R2022L XP_011511986.1
XM_011513685.2 6268 Intron XP_011511987.1
XM_017008046.1 6268 Missense Mutation CGC,CTC R2991L XP_016863535.1
Gene
SLC10A4
Gene Name
solute carrier family 10 member 4
There are no transcripts associated with this gene.

Gene
ZAR1
Gene Name
zygote arrest 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175619.2 6268 Intron NP_783318.1

View Full Product Details