Product Details

SNP ID
rs201521313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88467198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTATCATCAAGTGAAGAACTC[A/G]AACTTGGTAAATTCTATAGGAACAT
Phenotype
MIM: 608242
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HERC5 PubMed Links

Gene Details

Gene
HERC5
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016323.3 1717 Missense Mutation AAA,GAA K351E NP_057407.2
XM_011532022.2 1717 Missense Mutation AAA,GAA K502E XP_011530324.2

View Full Product Details