Product Details

SNP ID
rs201693237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158713256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATAACAGCCTTTGAACTGTCCA[C/T]GTATCTGCAGAATGCATTAATAAAA
Phenotype
MIM: 231675 MIM: 601753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETFDH PubMed Links

Gene Details

Gene
ETFDH
Gene Name
electron transfer flavoprotein dehydrogenase
There are no transcripts associated with this gene.

Gene
PPID
Gene Name
peptidylprolyl isomerase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005038.2 869 Missense Mutation ATG,GTG M253V NP_005029.1

View Full Product Details