Product Details

SNP ID
rs201179107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:42401112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTGTTGCACCTGTTTGAGACC[C/T]AAGGAGCCCTCGCAGCAGCCAATCC
Phenotype
MIM: 609542
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8A1 PubMed Links

Gene Details

Gene
ATP8A1
Gene Name
ATPase phospholipid transporting 8A1
There are no transcripts associated with this gene.

Gene
LOC105374428
Gene Name
uncharacterized LOC105374428
There are no transcripts associated with this gene.

Gene
SHISA3
Gene Name
shisa family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080505.1 596 Silent Mutation CCC,CCT P126P NP_001073974.1

View Full Product Details