Product Details

SNP ID
rs201592611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:72283167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAAGGTGGATGATCTTGTCGGA[C/T]GTCTGTTGTCAATGATCTTTCCATC
Phenotype
MIM: 605011
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS3 PubMed Links

Gene Details

Gene
ADAMTS3
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014243.2 4155 Missense Mutation CAT,CGT H1196R NP_055058.2
XM_011532421.2 4155 Missense Mutation CAT,CGT H1177R XP_011530723.1
XM_011532422.2 4155 Missense Mutation CAT,CGT H1168R XP_011530724.1

View Full Product Details