Product Details

SNP ID
rs202242697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1201747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCAGGAGGAGGCCAGCTCCCGG[A/C]CGAAGTGGGACAACAAGGCGCAGTA
Phenotype
MIM: 608893
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC6A19 PubMed Links

Gene Details

Gene
SLC6A19
Gene Name
solute carrier family 6 member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003841.2 153 Missense Mutation ACG,CCG T33P NP_001003841.1

View Full Product Details