Product Details

SNP ID
rs202204644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140668273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAGAGGGAGAAACCAAGGAGCT[C/G]TGGTCATCAGGTCACCATCTCAAGG
Phenotype
MIM: 609385 MIM: 142810 MIM: 600549
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DND1 PubMed Links

Gene Details

Gene
DND1
Gene Name
DND microRNA-mediated repression inhibitor 1
There are no transcripts associated with this gene.

Gene
HARS
Gene Name
histidyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
IK
Gene Name
IK cytokine, down-regulator of HLA II
There are no transcripts associated with this gene.

Gene
WDR55
Gene Name
WD repeat domain 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017706.4 345 Silent Mutation CTC,CTG L77L NP_060176.2
XM_005268469.2 345 Silent Mutation CTC,CTG L77L XP_005268526.1
XM_017009600.1 345 UTR 5 XP_016865089.1

View Full Product Details