Product Details

SNP ID
rs202245502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160405047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACTTACAACAATCTCCTTCCCA[C/G]CTTCTCCAGTACAATAGGAATACTT
Phenotype
MIM: 605974 MIM: 615253
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLU7 PubMed Links

Gene Details

Gene
SLU7
Gene Name
SLU7 homolog, splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006425.4 1878 Missense Mutation GCT,GGT A459G NP_006416.3
XM_005265793.3 1878 Missense Mutation GCT,GGT A469G XP_005265850.1
XM_011534418.2 1878 Missense Mutation GCT,GGT A459G XP_011532720.1
Gene
ZBED8
Gene Name
zinc finger BED-type containing 8
There are no transcripts associated with this gene.

View Full Product Details