Product Details

SNP ID
rs202189745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135029091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGCTGGGTGCTGAGAACATGGA[C/T]TGCGACGACAGCGGGCTCATGGAGT
Phenotype
MIM: 602149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf66 PubMed Links

Gene Details

Gene
C5orf66
Gene Name
chromosome 5 open reading frame 66
There are no transcripts associated with this gene.

Gene
C5orf66-AS1
Gene Name
C5orf66 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PITX1
Gene Name
paired like homeodomain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002653.4 1026 Silent Mutation CAA,CAG Q211Q NP_002644.4

View Full Product Details