Product Details
- SNP ID
-
rs202197693
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:153992986 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACGTCTCCATTCTTCAATGTTCT[A/G]ACAAAGGTTTCTGGCCCTGCAGCTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM114A2
PubMed Links
Gene Details
- Gene
- FAM114A2
- Gene Name
- family with sequence similarity 114 member A2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001317993.1 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
NP_001304922.1 |
NM_001317994.1 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
NP_001304923.1 |
NM_001317995.1 |
1604 |
Missense Mutation |
TCA,TTA |
S433L |
NP_001304924.1 |
NM_018691.3 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
NP_061161.2 |
XM_005268360.4 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
XP_005268417.1 |
XM_005268361.4 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
XP_005268418.1 |
XM_011537540.2 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
XP_011535842.1 |
XM_011537541.2 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
XP_011535843.1 |
XM_011537542.2 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
XP_011535844.1 |
XM_017008967.1 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
XP_016864456.1 |
XM_017008968.1 |
1604 |
Missense Mutation |
TCA,TTA |
S503L |
XP_016864457.1 |
View Full Product Details