Product Details
- SNP ID
-
rs200287957
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:26885667 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCAACGAGAGCTCCCGTGCTCAGG[A/C]CGGCTGAAAGGATCAGGGCTTCTGC
- Phenotype
-
MIM: 609974
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDH9
PubMed Links
Gene Details
- Gene
- CDH9
- Gene Name
- cadherin 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016279.3 |
1998 |
Missense Mutation |
GGC,GTC |
G610V |
NP_057363.3 |
View Full Product Details