Product Details

SNP ID
rs201036501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37064562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAAAACGAAATTCAGACTCTAC[A/G]GAGTTGGCAGCACAGATGAATGAAA
Phenotype
MIM: 614571 MIM: 608667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C5orf42 PubMed Links

Gene Details

Gene
C5orf42
Gene Name
chromosome 5 open reading frame 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023073.3 8375 Intron NP_075561.3
XM_005248345.3 8375 Intron XP_005248402.1
XM_005248346.3 8375 Intron XP_005248403.1
XM_005248347.3 8375 Intron XP_005248404.1
XM_005248349.3 8375 Intron XP_005248406.1
XM_005248350.3 8375 Intron XP_005248407.1
XM_006714491.3 8375 Intron XP_006714554.1
XM_011514085.2 8375 Intron XP_011512387.1
XM_011514086.2 8375 Intron XP_011512388.1
XM_011514087.2 8375 Intron XP_011512389.1
XM_011514088.2 8375 Intron XP_011512390.1
XM_011514089.2 8375 Intron XP_011512391.1
XM_011514090.2 8375 Intron XP_011512392.1
XM_011514092.2 8375 Intron XP_011512394.1
XM_011514094.2 8375 Intron XP_011512396.1
XM_017009760.1 8375 Intron XP_016865249.1
XM_017009761.1 8375 Intron XP_016865250.1
XM_017009762.1 8375 Intron XP_016865251.1
XM_017009763.1 8375 Intron XP_016865252.1
XM_017009764.1 8375 Intron XP_016865253.1
XM_017009765.1 8375 Intron XP_016865254.1
XM_017009766.1 8375 Intron XP_016865255.1
Gene
NIPBL
Gene Name
NIPBL, cohesin loading factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015384.4 8375 UTR 3 NP_056199.2
NM_133433.3 8375 Silent Mutation ACA,ACG T2695T NP_597677.2
XM_005248280.3 8375 UTR 3 XP_005248337.1
XM_005248282.4 8375 Silent Mutation ACA,ACG T2475T XP_005248339.3
XM_006714467.2 8375 Silent Mutation ACA,ACG T2646T XP_006714530.1
XM_006714468.2 8375 Silent Mutation ACA,ACG T2629T XP_006714531.1
XM_011514015.1 8375 Intron XP_011512317.1
XM_017009329.1 8375 UTR 3 XP_016864818.1
XM_017009330.1 8375 Silent Mutation ACA,ACG T2156T XP_016864819.1
XM_017009331.1 8375 Silent Mutation ACA,ACG T2153T XP_016864820.1

View Full Product Details