Product Details
- SNP ID
-
rs201036666
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:177524712 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGATCACCCGGGATAGGTTGGGC[A/G]CGGTGTTGAGTCTCCTCACGAGCCC
- Phenotype
-
MIM: 608170
MIM: 615813
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DDX41
PubMed Links
Gene Details
- Gene
- DDX41
- Gene Name
- DEAD-box helicase 41
There are no transcripts associated with this gene.
- Gene
- FAM193B
- Gene Name
- family with sequence similarity 193 member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001190946.2 |
2601 |
Missense Mutation |
GCG,GTG |
A590V |
NP_001177875.1 |
XM_006714875.2 |
2601 |
Missense Mutation |
GCG,GTG |
A670V |
XP_006714938.1 |
XM_006714876.1 |
2601 |
Missense Mutation |
GCG,GTG |
A557V |
XP_006714939.1 |
XM_006714878.1 |
2601 |
Missense Mutation |
GCG,GTG |
A557V |
XP_006714941.1 |
XM_006714879.3 |
2601 |
Missense Mutation |
GCG,GTG |
A557V |
XP_006714942.1 |
XM_006714880.1 |
2601 |
Missense Mutation |
GCG,GTG |
A557V |
XP_006714943.1 |
XM_006714884.1 |
2601 |
Missense Mutation |
GCG,GTG |
A216V |
XP_006714947.1 |
XM_006714885.1 |
2601 |
Missense Mutation |
GCG,GTG |
A216V |
XP_006714948.1 |
XM_006714886.2 |
2601 |
Intron |
|
|
XP_006714949.1 |
XM_011534572.1 |
2601 |
Missense Mutation |
GCG,GTG |
A635V |
XP_011532874.1 |
XM_011534573.1 |
2601 |
Missense Mutation |
GCG,GTG |
A626V |
XP_011532875.1 |
XM_011534574.1 |
2601 |
Missense Mutation |
GCG,GTG |
A592V |
XP_011532876.1 |
XM_011534579.2 |
2601 |
Missense Mutation |
GCG,GTG |
A565V |
XP_011532881.1 |
XM_011534580.1 |
2601 |
Missense Mutation |
GCG,GTG |
A557V |
XP_011532882.1 |
XM_011534581.1 |
2601 |
Missense Mutation |
GCG,GTG |
A354V |
XP_011532883.1 |
XM_011534582.1 |
2601 |
Missense Mutation |
GCG,GTG |
A216V |
XP_011532884.1 |
XM_011534583.1 |
2601 |
Missense Mutation |
GCG,GTG |
A216V |
XP_011532885.1 |
View Full Product Details