Product Details
- SNP ID
-
rs201178263
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:132074858 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGGAGCTGTACAAGCAGGGCCTG[C/T]GGGGCAGCCTCACCAAGCTCAAGGG
- Phenotype
-
MIM: 138960
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CSF2
PubMed Links
Gene Details
- Gene
- CSF2
- Gene Name
- colony stimulating factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000758.3 |
282 |
Missense Mutation |
CGG,TGG |
R84W |
NP_000749.2 |
View Full Product Details