Product Details

SNP ID
rs200549197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:181224650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGATGCACCCAACCCCTGGTCG[A/T]GGGAGCCGCGTGACCGATCAGGGCA
Phenotype
MIM: 610530
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR4638 PubMed Links

Gene Details

Gene
MIR4638
Gene Name
microRNA 4638
There are no transcripts associated with this gene.

Gene
TRIM41
Gene Name
tripartite motif containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033549.4 1386 Silent Mutation CGA,CGT R217R NP_291027.3
NM_201627.2 1386 Silent Mutation CGA,CGT R217R NP_963921.1
XM_006714929.2 1386 Silent Mutation CGA,CGT R217R XP_006714992.1
XM_006714930.2 1386 Silent Mutation CGA,CGT R217R XP_006714993.1
XM_006714931.3 1386 Silent Mutation CGA,CGT R217R XP_006714994.1
XM_006714932.3 1386 Silent Mutation CGA,CGT R217R XP_006714995.1

View Full Product Details