Product Details

SNP ID
rs201423535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55425277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGCAGAGTTGTATGAGAGTCCT[C/T]CTCTTTTCTTTCTTTAAAAGAAGTT
Phenotype
MIM: 607124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLPP1 PubMed Links

Gene Details

Gene
PLPP1
Gene Name
phospholipid phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003711.3 3900 Missense Mutation AAG,GAG K262E NP_003702.2
NM_176895.2 3900 Missense Mutation AAG,GAG K263E NP_795714.1
XM_006714724.2 3900 Missense Mutation AAG,GAG K199E XP_006714787.1
Gene
SKIV2L2
Gene Name
Ski2 like RNA helicase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015360.4 3900 UTR 3 NP_056175.3

View Full Product Details