Product Details

SNP ID
rs200735877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177135245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGAGCCACTTAATGGGTGTACT[A/G]TGCAGTTATCGACTGTCAGTGGAAC
Phenotype
MIM: 606681
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSD1 PubMed Links

Gene Details

Gene
NSD1
Gene Name
nuclear receptor binding SET domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022455.4 280 Missense Mutation ATG,GTG M48V NP_071900.2
NM_172349.2 280 Intron NP_758859.1

View Full Product Details