Product Details

SNP ID
rs200422364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:111731017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTTGTCTCATCGTTCTTCTTGCG[C/G]TTCACTTCCTTTGGGACAGGAAGTC
Phenotype
MIM: 607332
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NREP PubMed Links

Gene Details

Gene
NREP
Gene Name
neuronal regeneration related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142474.1 590 Missense Mutation AAC,AAG N71K NP_001135946.1
NM_001142475.1 590 Missense Mutation AAC,AAG N81K NP_001135947.1
NM_001142476.1 590 Missense Mutation AAC,AAG N37K NP_001135948.1
NM_001142477.1 590 Intron NP_001135949.1
NM_001142478.1 590 Intron NP_001135950.1
NM_001142479.1 590 Intron NP_001135951.1
NM_001142480.1 590 Intron NP_001135952.1
NM_001142481.1 590 Intron NP_001135953.1
NM_001142482.1 590 Intron NP_001135954.1
NM_001142483.1 590 Missense Mutation AAC,AAG N37K NP_001135955.1
NM_004772.2 590 Intron NP_004763.1
XM_006714732.1 590 Intron XP_006714795.1
XM_006714733.2 590 Missense Mutation AAC,AAG N56K XP_006714796.1
Gene
STARD4-AS1
Gene Name
STARD4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details