Product Details

SNP ID
rs201671590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33279850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCGTGGGTCCAGACAAATAAGCT[A/C]TGCAGGTACCTGGGGGTGTCACAGA
Phenotype
MIM: 603095 MIM: 605660 MIM: 180473 MIM: 603443 MIM: 611440
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
B3GALT4 PubMed Links

Gene Details

Gene
B3GALT4
Gene Name
beta-1,3-galactosyltransferase 4
There are no transcripts associated with this gene.

Gene
MIR6873
Gene Name
microRNA 6873
There are no transcripts associated with this gene.

Gene
PFDN6
Gene Name
prefoldin subunit 6
There are no transcripts associated with this gene.

Gene
RPS18
Gene Name
ribosomal protein S18
There are no transcripts associated with this gene.

Gene
VPS52
Gene Name
VPS52, GARP complex subunit
There are no transcripts associated with this gene.

Gene
WDR46
Gene Name
WD repeat domain 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164267.1 1605 Nonsense Mutation GAG,TAG E458* NP_001157739.1
NM_005452.5 1605 Nonsense Mutation GAG,TAG E512* NP_005443.3
XM_017011484.1 1605 Nonsense Mutation GAG,TAG E512* XP_016866973.1
XM_017011485.1 1605 Nonsense Mutation GAG,TAG E190* XP_016866974.1
XM_017011486.1 1605 Nonsense Mutation GAG,TAG E168* XP_016866975.1
XM_017011487.1 1605 Nonsense Mutation GAG,TAG E168* XP_016866976.1

View Full Product Details