Product Details

SNP ID
rs267608246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42965115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGAACGCGTAGCTGGGAGGCCC[G/A]GTCCTCATTTGCCCCCACAAACACC
Phenotype
MIM: 606628 MIM: 601498
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CNPY3-GNMT PubMed Links

Gene Details

Gene
CNPY3-GNMT
Gene Name
CNPY3-GNMT readthrough
There are no transcripts associated with this gene.

Gene
GNMT
Gene Name
glycine N-methyltransferase
There are no transcripts associated with this gene.

Gene
PEX6
Gene Name
peroxisomal biogenesis factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000287.3 2719 Silent Mutation NP_000278.3
NM_001316313.1 2719 Silent Mutation NP_001303242.1
XM_011514661.1 2719 Missense Mutation XP_011512963.1

View Full Product Details