Product Details

SNP ID
rs201898032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:136922643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATCGCCGTGCCCCCCTTCGGTC[G/T]GAGCCTGGCGCGCTCCTTCGCGGGG
Phenotype
MIM: 601757 MIM: 612519
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PEX7 PubMed Links

Gene Details

Gene
PEX7
Gene Name
peroxisomal biogenesis factor 7
There are no transcripts associated with this gene.

Gene
SLC35D3
Gene Name
solute carrier family 35 member D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008783.2 403 Missense Mutation CGG,CTG R72L NP_001008783.1

View Full Product Details