Product Details

SNP ID
rs200601840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33315123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCGCACTCGCTTCGTCGCCGCT[G/T]CCCCCGCCCACTCCGGGAGACTCTC
Phenotype
MIM: 603186 MIM: 601962 MIM: 611439
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DAXX PubMed Links

Gene Details

Gene
DAXX
Gene Name
death domain associated protein
There are no transcripts associated with this gene.

Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003190.4 1928 Intron NP_003181.3
NM_172208.2 1928 Intron NP_757345.2
NM_172209.2 1928 Intron NP_757346.2
XM_011514828.2 1928 Intron XP_011513130.1
XM_017011227.1 1928 Intron XP_016866716.1
Gene
ZBTB22
Gene Name
zinc finger and BTB domain containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145338.1 1928 Silent Mutation GGA,GGC G598G NP_001138810.1
NM_005453.4 1928 Silent Mutation GGA,GGC G598G NP_005444.4

View Full Product Details