Product Details

SNP ID
rs201560709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26234303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGGAGCTGCCTTCTTTGCCTTTG[A/T]AACCTTCGGCTTCCCCGACTTAGGC
Phenotype
MIM: 142210 MIM: 602813 MIM: 602824
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H1D PubMed Links

Gene Details

Gene
HIST1H1D
Gene Name
histone cluster 1, H1d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005320.2 686 Missense Mutation ACA,TCA T211S NP_005311.1
Gene
HIST1H3E
Gene Name
histone cluster 1, H3e
There are no transcripts associated with this gene.

Gene
HIST1H4F
Gene Name
histone cluster 1, H4f
There are no transcripts associated with this gene.

View Full Product Details