Product Details

SNP ID
rs200704036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33573839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATCTTCGTACCACAAACTGGCC[C/G]AACAGAACCACACCCAGAACCACCA
Phenotype
MIM: 600516 MIM: 609495
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BAK1 PubMed Links

Gene Details

Gene
BAK1
Gene Name
BCL2 antagonist/killer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001188.3 Intron NP_001179.1
XM_011514779.2 Intron XP_011513081.1
XM_011514780.1 Intron XP_011513082.1
Gene
GGNBP1
Gene Name
gametogenetin binding protein 1 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details