Product Details

SNP ID
rs200685500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33304200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAAGGCCTACGCTGTCCTCAA[A/G]GGAGGGCCCTGAAAGACCTGGCAGG
Phenotype
MIM: 602306 MIM: 601962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGL2 PubMed Links

Gene Details

Gene
RGL2
Gene Name
ral guanine nucleotide dissociation stimulator like 2
There are no transcripts associated with this gene.

Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003190.4 1441 Missense Mutation CTT,TTT L410F NP_003181.3
NM_172208.2 1441 Missense Mutation CTT,TTT L410F NP_757345.2
NM_172209.2 1441 Missense Mutation CTT,TTT L323F NP_757346.2
XM_011514828.2 1441 Intron XP_011513130.1
XM_017011227.1 1441 Missense Mutation CTT,TTT L410F XP_016866716.1

View Full Product Details