Product Details

SNP ID
rs201078014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3225798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCCCTTGGCCCAGTTATTCCC[A/G]GCTCCACTCTGGCCTGCCAGAGGGA
Phenotype
MIM: 612850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TUBB2B PubMed Links

Gene Details

Gene
TUBB2B
Gene Name
tubulin beta 2B class IIb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178012.4 482 Silent Mutation NP_821080.1

View Full Product Details