Product Details

SNP ID
rs201197923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35805959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCCTCTAGAGCCTTCAAGACT[G/T]CCATGTTCTTTGTGGCCTTGGGCAT
Phenotype
MIM: 120105 MIM: 609427
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLPS PubMed Links

Gene Details

Gene
CLPS
Gene Name
colipase
There are no transcripts associated with this gene.

Gene
LHFPL5
Gene Name
lipoma HMGIC fusion partner-like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182548.3 666 Missense Mutation GCC,TCC A97S NP_872354.1

View Full Product Details