Product Details

SNP ID
rs201219042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41043698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCACAGTCCACAACCCTGGTCT[C/G]GTAAGGCACACAGTGCTCAGTAGCA
Phenotype
MIM: 604797
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APOBEC2 PubMed Links

Gene Details

Gene
APOBEC2
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 2
There are no transcripts associated with this gene.

Gene
TSPO2
Gene Name
translocator protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010873.2 426 Silent Mutation CTC,CTG L105L NP_001010873.1
NM_001159726.1 426 Silent Mutation CTC,CTG L105L NP_001153198.1
XM_011514396.2 426 Silent Mutation CTC,CTG L105L XP_011512698.1
XM_011514397.2 426 Silent Mutation CTC,CTG L105L XP_011512699.1
Gene
UNC5CL
Gene Name
unc-5 family C-terminal like
There are no transcripts associated with this gene.

View Full Product Details