Product Details
- SNP ID
-
rs200808642
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:156778394 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCTCAGCCCGGCCCCGACATGGA[C/G]CAGCCGCAACATGGAGGCGCCAAGG
- Phenotype
-
MIM: 614556
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARID1B
PubMed Links
Gene Details
- Gene
- ARID1B
- Gene Name
- AT-rich interaction domain 1B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017519.2 |
540 |
Silent Mutation |
GAC,GAG |
D155E |
NP_059989.2 |
NM_020732.3 |
540 |
Silent Mutation |
GAC,GAG |
D155E |
NP_065783.3 |
XM_005267069.4 |
540 |
Silent Mutation |
GAC,GAG |
D155E |
XP_005267126.2 |
XM_011535984.2 |
540 |
Silent Mutation |
GAC,GAG |
D155E |
XP_011534286.2 |
XM_011535988.2 |
540 |
Intron |
|
|
XP_011534290.1 |
XM_017011103.1 |
540 |
Silent Mutation |
GAC,GAG |
D155E |
XP_016866592.1 |
XM_017011104.1 |
540 |
Silent Mutation |
GAC,GAG |
D155E |
XP_016866593.1 |
XM_017011105.1 |
540 |
Silent Mutation |
GAC,GAG |
D155E |
XP_016866594.1 |
XM_017011106.1 |
540 |
Silent Mutation |
GAC,GAG |
D155E |
XP_016866595.1 |
XM_017011107.1 |
540 |
Silent Mutation |
GAC,GAG |
D155E |
XP_016866596.1 |
XM_017011108.1 |
540 |
Missense Mutation |
GAC,GAG |
D182E |
XP_016866597.1 |
XM_017011109.1 |
540 |
Intron |
|
|
XP_016866598.1 |
- Gene
- MIR4466
- Gene Name
- microRNA 4466
There are no transcripts associated with this gene.
View Full Product Details