Product Details

SNP ID
rs201548947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3224798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCTCGAACTCCCCTTGTTCGT[C/T]GGCCGTGGCGTCCTGGTACTGCTGG
Phenotype
MIM: 612850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TUBB2B PubMed Links

Gene Details

Gene
TUBB2B
Gene Name
tubulin beta 2B class IIb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178012.4 1482 Missense Mutation AAC,GAC N431D NP_821080.1

View Full Product Details