Product Details
- SNP ID
-
rs200356862
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:149578901 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTAAGGATTTTAGTTCATGAGTG[A/G]CCTATGACATCTGGTTCCAGTTTGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GINM1
PubMed Links
Gene Details
- Gene
- GINM1
- Gene Name
- glycoprotein integral membrane 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138785.3 |
380 |
Missense Mutation |
TGA,TGG |
*119W |
NP_620140.1 |
View Full Product Details