Product Details

SNP ID
rs200795261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30651345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTTGCACACCTTCGTTTGGAG[A/G]TTTTACAGCTGACCCGCCACCCTGA
Phenotype
MIM: 615556 MIM: 603405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATAT1 PubMed Links

Gene Details

Gene
ATAT1
Gene Name
alpha tubulin acetyltransferase 1
There are no transcripts associated with this gene.

Gene
C6orf136
Gene Name
chromosome 6 open reading frame 136
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109938.2 434 Missense Mutation ATT,GTT I215V NP_001103408.1
NM_001161376.1 434 Missense Mutation ATT,GTT I396V NP_001154848.1
NM_145029.3 434 Missense Mutation ATT,GTT I81V NP_659466.2
XM_011514384.1 434 Missense Mutation ATT,GTT I72V XP_011512686.1
Gene
DHX16
Gene Name
DEAH-box helicase 16
There are no transcripts associated with this gene.

View Full Product Details