Product Details

SNP ID
rs201208600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42208905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCAGGTAAGTTTCGCTGAATAT[A/C]TTCCAAGTAGACATCTGCTGTGCTT
Phenotype
MIM: 611976
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRPS10 PubMed Links

Gene Details

Gene
MRPS10
Gene Name
mitochondrial ribosomal protein S10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018141.3 522 Missense Mutation GAT,TAT D159Y NP_060611.2
XM_005249206.2 522 Missense Mutation GAT,TAT D158Y XP_005249263.1
XM_011514724.2 522 Missense Mutation GAT,TAT D169Y XP_011513026.1
XM_011514725.2 522 Missense Mutation GAT,TAT D168Y XP_011513027.1
XM_017010996.1 522 Missense Mutation GAT,TAT D118Y XP_016866485.1

View Full Product Details