Product Details
- SNP ID
-
rs201251694
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:158663288 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGAAATGGCCCAAGAAATAGATC[A/T]GAGTGCTCTCAAGGAGTTAGAACGC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SYTL3
PubMed Links
Gene Details
- Gene
- SYTL3
- Gene Name
- synaptotagmin like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001009991.3 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
NP_001009991.2 |
NM_001242384.1 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
NP_001229313.1 |
NM_001242394.1 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
NP_001229323.1 |
NM_001242395.1 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
NP_001229324.1 |
NM_001318745.1 |
690 |
Intron |
|
|
NP_001305674.1 |
XM_005267215.4 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
XP_005267272.1 |
XM_005267218.4 |
690 |
Intron |
|
|
XP_005267275.1 |
XM_005267222.4 |
690 |
Intron |
|
|
XP_005267279.1 |
XM_006715605.3 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
XP_006715668.1 |
XM_006715606.3 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
XP_006715669.1 |
XM_006715611.3 |
690 |
Intron |
|
|
XP_006715674.1 |
XM_011536254.2 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
XP_011534556.1 |
XM_011536255.2 |
690 |
Intron |
|
|
XP_011534557.1 |
XM_017011495.1 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
XP_016866984.1 |
XM_017011496.1 |
690 |
UTR 5 |
|
|
XP_016866985.1 |
XM_017011497.1 |
690 |
UTR 5 |
|
|
XP_016866986.1 |
XM_017011498.1 |
690 |
Intron |
|
|
XP_016866987.1 |
XM_017011499.1 |
690 |
Missense Mutation |
CAG,CTG |
Q7L |
XP_016866988.1 |
View Full Product Details