Product Details

SNP ID
rs199870664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:148747440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCGGAGCAGGCGATGAAGAAG[A/C]AGCAGCAGCATCCCGGCGGCGGCGC
Phenotype
MIM: 610752
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UST PubMed Links

Gene Details

Gene
UST
Gene Name
uronyl 2-sulfotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005715.2 997 Missense Mutation AAG,CAG K4Q NP_005706.1
XM_011535378.2 997 Missense Mutation AAG,CAG K4Q XP_011533680.1
XM_017010152.1 997 Missense Mutation AAG,CAG K4Q XP_016865641.1

View Full Product Details