Product Details

SNP ID
rs199474438
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29942955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGGGACCAGGAGACACGGAATG[C/T]GAAGGCCCAGTCACAGACTGACCGA
Phenotype
MIM: 142800
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HLA-A PubMed Links

Gene Details

Gene
HLA-A
Gene Name
major histocompatibility complex, class I, A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242758.1 356 Missense Mutation ACG,ATG T91M NP_001229687.1
NM_002116.7 356 Missense Mutation GCG,GTG A91V NP_002107.3

View Full Product Details