Product Details

SNP ID
rs200212980
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26033407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGGCGGTCAGGTACTCAAGCAC[C/T]GCCGCGAGATACACCGGCGCGCCAG
Phenotype
MIM: 602795 MIM: 602803 MIM: 602819 MIM: 602829
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HIST1H2AB PubMed Links
Additional Information
For this assay, SNP(s) [rs2230654] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HIST1H2AB
Gene Name
histone cluster 1, H2ab
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003513.2 Intron NP_003504.2
Gene
HIST1H2BB
Gene Name
histone cluster 1, H2bb
There are no transcripts associated with this gene.

Gene
HIST1H3B
Gene Name
histone cluster 1, H3b
There are no transcripts associated with this gene.

Gene
HIST1H4B
Gene Name
histone cluster 1, H4b
There are no transcripts associated with this gene.

View Full Product Details