Product Details
- SNP ID
-
rs199585067
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:150025409 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCGTCTCGCCTAGCCCGGGACCA[A/G]CCGAACAGCAGGAACAGAAGCGGGA
- Phenotype
-
MIM: 611047
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RAET1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117766916,rs1543547] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RAET1L
- Gene Name
- retinoic acid early transcript 1L
View Full Product Details