Product Details

SNP ID
rs199723551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27249170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGAACACAGATTTTATGGCCT[G/T]AGTATACCTGCTGGAGGCCTGGAAA
Phenotype
MIM: 607169
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRSS16 PubMed Links

Gene Details

Gene
PRSS16
Gene Name
protease, serine 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005865.3 423 Silent Mutation CTG,CTT L136L NP_005856.1
XM_017010161.1 423 Silent Mutation CTG,CTT L136L XP_016865650.1
XM_017010162.1 423 Silent Mutation CTG,CTT L136L XP_016865651.1
XM_017010163.1 423 Silent Mutation CTG,CTT L136L XP_016865652.1
XM_017010164.1 423 Silent Mutation CTG,CTT L136L XP_016865653.1
XM_017010165.1 423 Intron XP_016865654.1

View Full Product Details