Product Details
- SNP ID
-
rs199803961
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:53005178 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCGAGATGCGTACTTGGAAGCCC[A/C]GTCTGTCCGGCCATGCACTGGCTGG
- Phenotype
-
MIM: 605450
MIM: 612325
MIM: 606515
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GSTA4
PubMed Links
Gene Details
- Gene
- GSTA4
- Gene Name
- glutathione S-transferase alpha 4
There are no transcripts associated with this gene.
- Gene
- ICK
- Gene Name
- intestinal cell (MAK-like) kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014920.3 |
3050 |
Missense Mutation |
GGG,TGG |
G624W |
NP_055735.1 |
NM_016513.4 |
3050 |
Missense Mutation |
GGG,TGG |
G624W |
NP_057597.2 |
XM_011514419.1 |
3050 |
Missense Mutation |
GGG,TGG |
G631W |
XP_011512721.1 |
XM_011514420.2 |
3050 |
Missense Mutation |
GGG,TGG |
G631W |
XP_011512722.1 |
XM_011514421.1 |
3050 |
Missense Mutation |
GGG,TGG |
G631W |
XP_011512723.1 |
XM_017010485.1 |
3050 |
Missense Mutation |
GGG,TGG |
G631W |
XP_016865974.1 |
XM_017010486.1 |
3050 |
Missense Mutation |
GGG,TGG |
G631W |
XP_016865975.1 |
XM_017010487.1 |
3050 |
Missense Mutation |
GGG,TGG |
G631W |
XP_016865976.1 |
XM_017010488.1 |
3050 |
Missense Mutation |
GGG,TGG |
G624W |
XP_016865977.1 |
XM_017010489.1 |
3050 |
Missense Mutation |
GGG,TGG |
G624W |
XP_016865978.1 |
XM_017010490.1 |
3050 |
Missense Mutation |
GGG,TGG |
G624W |
XP_016865979.1 |
XM_017010491.1 |
3050 |
Missense Mutation |
GGG,TGG |
G624W |
XP_016865980.1 |
XM_017010492.1 |
3050 |
Missense Mutation |
GGG,TGG |
G624W |
XP_016865981.1 |
- Gene
- RN7SK
- Gene Name
- RNA, 7SK small nuclear
There are no transcripts associated with this gene.
View Full Product Details