Product Details

SNP ID
rs200944951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87402096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAAATATACTGTAGCAAAAGT[C/T]CATAAGTTCTGTGTCCCAGCCTGGA
Phenotype
MIM: 171060 MIM: 606090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCB4 PubMed Links

Gene Details

Gene
ABCB4
Gene Name
ATP binding cassette subfamily B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011516308.2 Intron XP_011514610.2
XM_011516309.2 Intron XP_011514611.2
XM_011516310.2 Intron XP_011514612.2
XM_011516311.2 Intron XP_011514613.2
XM_011516312.2 Intron XP_011514614.2
XM_011516315.2 Intron XP_011514617.2
XM_017012323.1 Intron XP_016867812.1
Gene
CROT
Gene Name
carnitine O-octanoyltransferase
There are no transcripts associated with this gene.

View Full Product Details