Product Details

SNP ID
rs200567913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:18027352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGACAGTTGGCTCTGCATACAAG[C/T]TGTCTACTGGGATATCAAAAAAGCC
Phenotype
MIM: 611566
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPS1L1 PubMed Links

Gene Details

Gene
PRPS1L1
Gene Name
phosphoribosyl pyrophosphate synthetase 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175886.2 512 Missense Mutation AAC,AGC N144S NP_787082.1

View Full Product Details