Product Details

SNP ID
rs199875419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27182997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCGGATCTGGTACTTGGTATA[C/G]GGGCAGCGCTTTTTGCGGGTGCGTT
Phenotype
MIM: 142957 MIM: 142958 MIM: 607530
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HOXA10 PubMed Links

Gene Details

Gene
HOXA10
Gene Name
homeobox A10
There are no transcripts associated with this gene.

Gene
HOXA10-HOXA9
Gene Name
HOXA10-HOXA9 readthrough
There are no transcripts associated with this gene.

Gene
HOXA11
Gene Name
homeobox A11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005523.5 813 Silent Mutation CCC,CCG P247P NP_005514.1
Gene
HOXA11-AS
Gene Name
HOXA11 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details