Product Details
- SNP ID
-
rs200380921
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:143183423 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCAAGAGTCTGCCCATCAAATGC[G/T]AGGGAGATGTTTTCCTCCAGACACC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TAS2R39
PubMed Links
Gene Details
- Gene
- TAS2R39
- Gene Name
- taste 2 receptor member 39
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_176881.2 |
5 |
Missense Mutation |
CGA,CTA |
R2L |
NP_795362.2 |
View Full Product Details