Product Details

SNP ID
rs200967332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150339049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAGAACTTTGTCCTCAGAGCC[C/T]AGTTTGATGCAGGCCAGGCATTTCC
Phenotype
MIM: 601973 MIM: 615252
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC61 PubMed Links

Gene Details

Gene
LRRC61
Gene Name
leucine rich repeat containing 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142928.1 429 Intron NP_001136400.1
NM_023942.2 429 Intron NP_076431.1
XM_006716095.2 429 Intron XP_006716158.1
Gene
RARRES2
Gene Name
retinoic acid receptor responder 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002889.3 429 Silent Mutation CTA,CTG L104L NP_002880.1
XM_017012491.1 429 Intron XP_016867980.1
Gene
ZBED6CL
Gene Name
ZBED6 C-terminal like
There are no transcripts associated with this gene.

View Full Product Details