Product Details
- SNP ID
-
hCV191775151
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:73570951 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTGTCCCACAGTTTCCATGTACC[A/G]TCCTTGGAGACAGAAGCCATCCTGC
- Phenotype
-
MIM: 605842
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TBL2
PubMed Links
Gene Details
- Gene
- TBL2
- Gene Name
- transducin (beta)-like 2
There are no transcripts associated with this gene.
View Full Product Details