Product Details

SNP ID
rs200557896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149250246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGCTCCTATTTTCAGACCACC[A/G]AGATTTCACTCTGGACGGTGGTGGC
Phenotype
MIM: 602386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF212 PubMed Links

Gene Details

Gene
ZNF212
Gene Name
zinc finger protein 212
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012256.3 238 Missense Mutation AAG,GAG K38E NP_036388.2
XM_005250055.2 238 Missense Mutation AAG,GAG K78E XP_005250112.2
XM_011516581.2 238 UTR 5 XP_011514883.2
XM_011516582.1 238 UTR 5 XP_011514884.1
XM_017012646.1 238 Missense Mutation AAG,GAG K38E XP_016868135.1
XM_017012647.1 238 Missense Mutation AAG,GAG K35E XP_016868136.1
XM_017012648.1 238 UTR 5 XP_016868137.1

View Full Product Details