Product Details
- SNP ID
-
rs200807787
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:117278164 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGACGCCTGCTGGGGTCATGTAGC[A/G]GTTGTCCAGTCAGCGTTCTTGGGGG
- Phenotype
-
MIM: 147870
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WNT2
PubMed Links
Gene Details
- Gene
- WNT2
- Gene Name
- Wnt family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003391.2 |
1374 |
Silent Mutation |
ACC,ACT |
T358T |
NP_003382.1 |
View Full Product Details